![Camphor United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/248/190/5c7d9c16-4aea-4918-8517-6060bab2e5ff/640/5c7d9c16-4aea-4918-8517-6060bab2e5ff.png)
Camphor
![Camphor United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/248/190/5c7d9c16-4aea-4918-8517-6060bab2e5ff/640/5c7d9c16-4aea-4918-8517-6060bab2e5ff.png)
Synonym(s):
D-Camphor, 2-Bornanone, 2-Camphanone
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing