![Calcitonin Gene Related Peptide human ≥95% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/images/363/571/76ca2e1b-8bb3-4365-90d3-47ec0dcb81b2/640/76ca2e1b-8bb3-4365-90d3-47ec0dcb81b2.jpg)
Calcitonin Gene Related Peptide human
![Calcitonin Gene Related Peptide human ≥95% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/images/363/571/76ca2e1b-8bb3-4365-90d3-47ec0dcb81b2/640/76ca2e1b-8bb3-4365-90d3-47ec0dcb81b2.jpg)
Synonym(s):
α-CGRP, CGRP-I
Empirical Formula (Hill Notation):
C163H267N51O49S2
CAS Number:
Molecular Weight:
3789.31
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing