
Bromodichloroacetic acid

Synonym(s):
Bromdichloroacetic acid
Linear Formula:
BrCl2CCOOH
CAS Number:
Molecular Weight:
207.84
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1