Skip to Content
MilliporeSigma
Benzyl acetate analytical standard

Benzyl acetate

Benzyl acetate analytical standard
Synonym(s):
Acetic acid benzyl ester, Benzyl acetate
Linear Formula:
CH3COOCH2C6H5
CAS Number:
Molecular Weight:
150.17
EC Number:

Products

Shipping
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing