ar-Turmerone
Synonym(s):
(S)-ar-Turmerone, (S)-2-Methyl-6-(4-methylphenyl)-2-hepten-4-one
Empirical Formula (Hill Notation):
C15H20O
CAS Number:
Molecular Weight:
216.32
Products
Formula Weight
Manufacturer
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing