
Anti-CD90.2 Antibody (mouse), PE-Cy7, clone 30-H12

Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2