
all-cis-7,10,13,16-Docosatetraenoic acid

Synonym(s):
cis-7,10,13,16-Docosatetraenoic acid, DTA
Empirical Formula (Hill Notation):
C22H36O2
CAS Number:
Molecular Weight:
332.52
Products
Brand
Formula Weight
Showing 1-1 of 1
Compare
Product Number
Description
Pricing