Skip to Content
MilliporeSigma
Adenosine 5′-diphosphate bis(cyclohexylammonium) salt ≥95% (HPLC), crystalline, bacterial

Adenosine 5′-diphosphate bis(cyclohexylammonium) salt

Adenosine 5′-diphosphate bis(cyclohexylammonium) salt ≥95% (HPLC), crystalline, bacterial
Synonym(s):
ADP
Empirical Formula (Hill Notation):
C10H15N5O10P2 · 2C6H13N
CAS Number:
Molecular Weight:
625.55

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing