Skip to Content
MilliporeSigma
Adamantane-d16 ≥98 atom % D, ≥98% (CP)

Adamantane-d16

Adamantane-d16 ≥98 atom % D, ≥98% (CP)
Empirical Formula (Hill Notation):
C10D16
CAS Number:
Molecular Weight:
152.33

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing