Skip to Content
MilliporeSigma
5-Keto-D-gluconic acid potassium salt ≥98.0% (TLC)

5-Keto-D-gluconic acid potassium salt

5-Keto-D-gluconic acid potassium salt ≥98.0% (TLC)
Synonym(s):
D-Xylo-5-hexulofuranosonic acid potassium salt
Empirical Formula (Hill Notation):
C6H9KO7
CAS Number:
Molecular Weight:
232.23

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing