![5-Bromo-6-chloro-pyrazin-2-amine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/351/549/b853c440-b515-46dc-9560-5f00170bcc73/640/b853c440-b515-46dc-9560-5f00170bcc73.png)
5-Bromo-6-chloro-pyrazin-2-amine
![5-Bromo-6-chloro-pyrazin-2-amine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/351/549/b853c440-b515-46dc-9560-5f00170bcc73/640/b853c440-b515-46dc-9560-5f00170bcc73.png)
Empirical Formula (Hill Notation):
C4H3BrClN3
CAS Number:
Molecular Weight:
208.44
Products
Product Category
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing