
5-Amino-3-ethyl-1H-pyrazole

Synonym(s):
3-Amino-5-ethyl-2H-pyrazole
Empirical Formula (Hill Notation):
C5H9N3
CAS Number:
Molecular Weight:
111.15
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1