Skip to Content
MilliporeSigma
4-Nitrophenyl 4-guanidinobenzoate hydrochloride protease inhibitor and substrate

4-Nitrophenyl 4-guanidinobenzoate hydrochloride

4-Nitrophenyl 4-guanidinobenzoate hydrochloride protease inhibitor and substrate
Synonym(s):
4-Guanidinobenzoic acid 4-nitrophenylester hydrochloride, NPGB, pNPGB
Empirical Formula (Hill Notation):
C14H12N4O4 · HCl
CAS Number:
Molecular Weight:
336.73
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing