Skip to Content
MilliporeSigma
4-Ethynylanisole 97%

4-Ethynylanisole

4-Ethynylanisole 97%
Synonym(s):
1-Ethynyl-4-methoxybenzene
Linear Formula:
CH3OC6H4C≡CH
CAS Number:
Molecular Weight:
132.16

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing