![3,4,9,10-Perylenetetracarboxylic dianhydride](/deepweb/assets/sigmaaldrich/product/structures/217/269/ce68f909-ee73-42be-88f5-9add8d2f170f/640/ce68f909-ee73-42be-88f5-9add8d2f170f.png)
3,4,9,10-Perylenetetracarboxylic dianhydride
![3,4,9,10-Perylenetetracarboxylic dianhydride](/deepweb/assets/sigmaaldrich/product/structures/217/269/ce68f909-ee73-42be-88f5-9add8d2f170f/640/ce68f909-ee73-42be-88f5-9add8d2f170f.png)
Synonym(s):
PTCDA, Perylene-3,4,9,10-tetracarboxylic dianhydride, Pigment Red 224
Empirical Formula (Hill Notation):
C24H8O6
CAS Number:
Molecular Weight:
392.32
Products
Shipping
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing