Skip to Content
MilliporeSigma
2,6-Dimethylphenol PESTANAL®, analytical standard

2,6-Dimethylphenol

2,6-Dimethylphenol PESTANAL®, analytical standard
Synonym(s):
2-Hydroxy-m-xylene, vic.-m-Xylenol, 2,6-Dimethylphenol
Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing