Skip to Content
MilliporeSigma
2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid Buffer substance HEPES EMPROVE® EXPERT

2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid

2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid Buffer substance HEPES EMPROVE® EXPERT
Synonym(s):
HEPES, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)
Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30

Products

Product Category
Brand
Formula Weight
pH Value
pK Value
Product Line
Quality Segment
Technique
Showing 1-1 of 1