![2-(3,3-Diethyl-1-triazenyl)-4-methylbenzenesulfonyl chloride](/deepweb/assets/sigmaaldrich/product/structures/425/304/bba23eea-0155-4e98-8521-67c6787fc927/640/bba23eea-0155-4e98-8521-67c6787fc927.png)
2-(3,3-Diethyl-1-triazenyl)-4-methylbenzenesulfonyl chloride
![2-(3,3-Diethyl-1-triazenyl)-4-methylbenzenesulfonyl chloride](/deepweb/assets/sigmaaldrich/product/structures/425/304/bba23eea-0155-4e98-8521-67c6787fc927/640/bba23eea-0155-4e98-8521-67c6787fc927.png)
Synonym(s):
2-[(1E)-3,3-diethyl-1-triazen-1-yl]-4-methyl-benzenesulfonyl chloride
Empirical Formula (Hill Notation):
C11H16ClN3O2S
CAS Number:
Molecular Weight:
289.78
Products
Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing