
2,2′-(o-Phenylenedioxy)diethanol

Synonym(s):
1,2-Bis(2-hydroxyethoxy)benzene
Linear Formula:
C6H4(OCH2CH2OH)2
CAS Number:
Molecular Weight:
198.22
Products
Shipping
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1