Skip to Content
MilliporeSigma
2,2′:5′,2′′-Terthiophene 99%

2,2′:5′,2′′-Terthiophene

2,2′:5′,2′′-Terthiophene 99%
Synonym(s):
α-Terthienyl, 2,5-Di(2-thienyl)thiophene
Empirical Formula (Hill Notation):
C12H8S3
CAS Number:
Molecular Weight:
248.39

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing