Skip to Content
MilliporeSigma
(1R)-(−)-Menthyl acetate analytical standard

(1R)-(−)-Menthyl acetate

(1R)-(−)-Menthyl acetate analytical standard
Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing