Skip to Content
MilliporeSigma
1,3,5-Tri[(3-pyridyl)-phen-3-yl]benzene ≥99% (HPLC)

1,3,5-Tri[(3-pyridyl)-phen-3-yl]benzene

1,3,5-Tri[(3-pyridyl)-phen-3-yl]benzene ≥99% (HPLC)
Synonym(s):
TmPyPB
Empirical Formula (Hill Notation):
C39H27N3
CAS Number:
Molecular Weight:
537.65

Products

Shipping
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing