Skip to Content
MilliporeSigma
1-[(2RS)-2-[[(4-chlorophenyl)methyl]oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole

1-[(2RS)-2-[[(4-chlorophenyl)methyl]oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole

1-[(2RS)-2-[[(4-chlorophenyl)methyl]oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole
Synonym(s):
Econazole
Empirical Formula (Hill Notation):
C18H15Cl3N2O
CAS Number:
Molecular Weight:
381.68

Products

Shipping
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1