Skip to Content
MilliporeSigma
1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanol

1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
α-(2,4-Difluorophenyl)-1H-1,2,4-triazole-1-ethanol
Empirical Formula (Hill Notation):
C10H9F2N3O
CAS Number:
Molecular Weight:
225.19

Products

Brand
Formula Weight
Manufacturer
Quality Segment
Special Grade
Showing 1-1 of 1