Product Name
RPMI-1640 Medium 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Product Number
r1145
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH1.8 - 2.2
Osmolality- mOs/kg
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
SterilityPass
Endotoxin Level<= 1 EU/ml
Glucose Concentration18.0 - 22.0 g/l