Product Name
Peripheral Blood Medium liquid, application tested
Product Number
p2602
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCECLEAR LIGHT RED SOLUTION
PH TEST AT RT7.1 - 7.5