Product Name
6-(7-Nitrobenzofurazan-4-ylamino)hexanoic acid suitable for fluorescence
Product Number
72964
Product Brand
sigma
CAS Number
88235-25-0
Molecular Weight
294.26
Test
Specification
APPEARANCE (COLOR)Faint Orange to Dark Orange and Faint Brown to Dark Brown
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 96.0 %
CARBON CONTENT48.98 % (THEORY)
HYDROGEN CONTENT4.80 % (THEORY)
NITROGEN CONTENT19.04 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM530.0 - 540.0 nm