Product Name
Methylene Blue solution concentrate according to Ehrlich, concentrated, aqueous solution
Product Number
03978
Product Brand
sigma
CAS Number
61-73-4
Molecular Weight
319.85
Test
Specification
APPEARANCE (COLOR)Blue to Very Dark Blue
APPEARANCE (FORM)Liquid
THIN LAYER CHROMATOGR.CORRESPONDS TO STANDARD
DENSITY D20/40.850 - 1.140
WAVELENGTH (1) (UV)656 - 666 nm
EXTINCTION (1%/1CM) (1) (UV)Report Result
SOLVENT (UV)Water
RECOMMENDED RETEST PERIOD30 MONTHS