Product Name
Dibenzothiophene analytical standard
Product Number
45776
Product Brand
sial
CAS Number
132-65-0
Molecular Weight
184.26
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and Faint Beige to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (GC AREA %)≥ 98.0 %
MELTING POINT96 - 102 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS