Product Name
Aflatoxin B1 solution 2 μg/mL in acetonitrile, analytical standard
Product Number
34029
Product Brand
sial
CAS Number
1162-65-8
Molecular Weight
312.27
Test
Specification
CONCENTRATION COMP1~ 2 ug/ml
MEASURING TOLERANCE P±≤ 0.200 ug/ml
SOLVENTACETONITRILE
PURITY (HPLC) COMP1≥ 98.0 %
MEASURING TOLERANCE P± COMP1≤ 2.0 %