Product Name
Acesulfame K for food analysis, ≥99.0%
Product Number
04054
Product Brand
sial
CAS Number
55589-62-3
Molecular Weight
201.24
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
ASSAY (HPLC WEIGHT %)≥ 99.0 %
CARBON CONTENT23.87 % (THEORY)
HYDROGEN CONTENT2.00 % (THEORY)
NITROGEN CONTENT6.96 % (THEORY)
SULFUR CONTENT15.93 % (Theory)
INFRARED SPECTRUMCONFORMS TO STRUCTURE