Product Name
3-Bromophenethylamine 97%
Product Number
534080
Product Brand
aldrich
CAS Number
58971-11-2
Molecular Weight
200.08
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
TITRATION96.5 - 103.5 %
TITRATION (METHOD)BROMINE COMBUSTION
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE