G004697
HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND I 2000 DMP, Ethanol:Hexane Mobile Phase
application for HPLC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
technique(s)
HPLC: suitable
test parameters
column: CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase: [A] ethanol; [B] hexane; (20:80, A:B)
flow rate: 1 mL/min
column temp.: 25 °C
detector: UV, 254 nm
injection: 1 μL
sample: 1 mg/mL in methanol
suitability
application for HPLC
application(s)
vitamins, nutraceuticals, and natural products
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
Analyte
Description
Flavanone, 98%
Analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service