Skip to Content
MilliporeSigma
All Photos(1)

Documents

ES5479

ChromegaChiral Chiral CCJ, 3 Micron HPLC Column

3 μm particle size, L × I.D. 15 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

material

stainless steel column

Quality Level

product line

ChromegaChiral

manufacturer/tradename

ES Industries 135151-CCJ

packaging

1 ea of

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 4.6 mm

matrix

fully porous particle

matrix active group

cellulose 4-methylbenzoate phase

particle size

3 μm

pore size

1000 Å pore size

operating pH

2-8

separation technique

chiral
supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

ChromegaChiral CCJ (cellulose 4-methylbenzoate) is a new product for high resolution chiral separations based on a new halogenated carbohydrate based chiral stationary phase.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChromegaChiral is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service