Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM78313

Supelco

Salicylaldehyde

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-Hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein/REAXYS Number:
471388
MDL number:
UNSPSC Code:
12352114
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

4.2 (vs air)

vapor pressure

1 mmHg ( 33 °C)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.573 (lit.)

bp

197 °C (lit.)

mp

1-2 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Oc1ccccc1C=O

InChI

1S/C7H6O2/c8-5-6-3-1-2-4-7(6)9/h1-5,9H

InChI key

SMQUZDBALVYZAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

170.6 °F - closed cup

flash_point_c

77 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service