Documents
54632
Rheodyne® Model 9725i PEEK Injector
About This Item
UNSPSC Code:
41115854
material
PEEK injector
description
manual; with MBB and switch
feature
manual
number of ports 6
positions 2
manufacturer/tradename
Rheodyne 9725I
parameter
5000 psi max. pressure
fitting
10-32 Rheodyne thread
General description
The 9725i contains a switch and has the make before break (MBB). With the MBB, flow is not interupted when switching from laod to inject.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)