Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SPR-488E

StressMarq Biosciences Inc

StressMarq Amyloid Beta 1-42 Oligomers

100 µg, pack of 5

Synonym(s):

Abeta Oligomers, Abeta peptide, Amyloid beta peptide oligomers, APP

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
4.5
PDF Icon
Shipped & Sold By StressMarq Biosciences Inc.
A Trusted Marketplace Seller
PDF Icon
Have a question?

assay

95

concentration

2 mg/mL

application(s)

life science and biopharma

biological source

human

mol wt

predicted mol wt 4.5 kDa

pH

7.4

storage temp.

-80 °C

storage condition

avoid repeated freeze/thaw cycles

manufacturer/tradename

StressMarq Biosciences Inc

Related Categories

General description

Human Synthetic Amyloid Beta 1-42 Oligomers are certified >95% pure using mass spec and HPLC.  They have been determined to be low endotoxin <2.5 EU/mL @ 1mg/mL. Validated for WB | In vivo Assay | In vitro Assay. Monomer source is catalog# SPR-485.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library