Documents
Z692735
microDOC Compact Gel Documentation System, with UV Transilluminator (UVT365)
AC/DC input 110 V AC
About This Item
UNSPSC Code:
41105300
manufacturer/tradename
Cleaver CSL-MDOCUV365$
General description
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 365 nm tranilluminator.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
Legal Information
TotalLab is a trademark of Nonlinear Dynamics Ltd.