Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z614025

Vivaspin 500 centrifugal concentrators

MWCO 10 kDa

Synonym(s):

500 µL Vivaspin, Centrifuges, Vivaspin Centrifugal Concentrator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

material

polycarbonate housing
polyethersulfone membrane

description

PES membrane

sterility

non-sterile

feature

holdup volume 5 μL

packaging

package of 25 ea

manufacturer/tradename

Sartorius VS0101

parameter

15000 × g max. centrifugal force

dead stop volume

5 μL

membrane area

0.5 cm2

tube size

~2.2 mL

working volume

500 μL

pore size

10 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.

Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service