Key Documents
TMO3664-0025
Nalgene® economy graduated cylinder
polypropylene, volume 25 mL, case of 18 ea
Synonym(s):
PP graduated cylinder, PP graduated flask
Select a Size
18 EACH
$125.00
$125.00
Estimated to ship onApril 19, 2025
Select a Size
Change View
18 EACH
$125.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$125.00
Estimated to ship onApril 19, 2025
material
polypropylene
packaging
case of 18 ea
manufacturer/tradename
Nalgene 3664-0025
capacity
25 mL
volume
25 mL
General description
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)