Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB4300106

Sigma-Aldrich

Anti-phospho-CHUK (pThr23) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IKBKA antibody produced in rabbit, Anti-IKK-alpha antibody produced in rabbit, Anti-IKK1 antibody produced in rabbit, Anti-IKKA antibody produced in rabbit, Anti-conserved helix-loop-helix ubiquitous kinase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~85 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(L-G-TP-G-G)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr23)

Gene Information

human ... CHUK(1147)

Immunogen

Peptide sequence around phosphorylation site of threonine 23 (L-G-T(p)-G-G), according to the protein CHUK.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Acts as part of the IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B thus leading to the dissociation of the inhibitor/NF-kappa-B complex and ultimately the degradation of the inhibitor. As part of the non-canonical pathway of NF-kappa-B activation, the MAP3K14-activated CHUK/IKKA homodimer phosphorylates NFKB2/p100 associated with RelB, inducing its proteolytic processing to NFKB2/p52 and the formation of NF-kappa-B RelB-p52 complexes. Also phosphorylates NCOA3. Phosphorylates 'Ser-10' of histone H3 at NF-kappa-B-regulated promoters during inflammatory responses triggered by cytokines.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Fang et al.
Cell death and differentiation, 23(9), 1471-1482 (2016-04-09)
Radioresistance is a major obstacle in successful clinical cancer radiotherapy, and the underlying mechanisms are not clear. Here we show that IKKα-mediated miR-196a biogenesis via interaction with Drosha regulates the sensitivity of nasopharyngeal carcinoma (NPC) cells to radiotherapy. Phosphorylation of
Bo Cen et al.
Gastroenterology, 158(4), 971-984 (2019-11-18)
Prostaglandin E2 (PGE2) promotes colorectal tumor formation and progression by unknown mechanisms. We sought to identify microRNAs (miRNAs) that might mediate the effects of PGE2 on colorectal cancer (CRC) development. We incubated LS174T colorectal cancer cells with PGE2 or without
Christin Elßner et al.
Gastroenterology, 156(4), 1190-1205 (2018-11-18)
Cholangiocyte proliferation and ductular reaction contribute to the onset and progression of liver diseases. Little is known about the role of the transcription factor nuclear factor-κB (NF-κB) in this process. We investigated the activities of the RELB proto-oncogene NF-κB subunit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service