Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB1407919

Sigma-Aldrich

Anti-C8ORF33 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ20989

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~25.3 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C8orf33(65265)

General description

The gene encoding chromosome 8 open reading frame 33 (C8orf33) is localized on human chromosome 8q24.

Immunogen

C8orf33 (AAH10001, 1 a.a. ~ 229 a.a) full-length human protein.

Sequence
MAALGHLAGEAAAAPGPGTPCASRGARLPGPVSSARNPSTVCLCPEQPTCSNADSRAHPLGDEGGTASKKQKNKKKTRNRASVANGGEKASEKLAPEEVPLSAEAQAQQLAQELAWCVEQLELGLKRQKPTPKQKEQAIGAIRTLRSKRTPLPRKRQLMHSLFGDYRAQMEAEWREALRALRAAAYSAQVQPVDGATRKKSQRVCRPRSIWRAKATLDMPDEEFRFNFF

Biochem/physiol Actions

Studies have shown that chromosome 8 open reading frame 33 (C8orf33) may have a role in G protein-coupled receptor signaling pathway, Ca2+ signaling and actin cytoskeleton modulation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiyong Xu et al.
Molecular cytogenetics, 7(1), 84-84 (2014-12-23)
The aims of this study were to evaluate the clinical utility of multiplex ligation-dependent probe amplification (MLPA) and array comparative genomic hybridization (aCGH) analyses on prenatal cases and to review prenatal ultrasound findings of cytogenomic syndromes. Of the 54 prenatal
The 8th annual pharmacogenetics in psychiatry meeting report.
K Aitchison et al.
The pharmacogenomics journal, 9(6), 358-361 (2009-10-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service