Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1305433

Sigma-Aldrich

ANTI-VIMENTIN (CENTER) antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

VIM, Vimentin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53652 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:10-1:50
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Li et al.
Molecular medicine reports, 19(6), 4727-4734 (2019-05-07)
The aim of the present study was to investigate the protective effect of integrin β1 in the treatment of stress urinary incontinence (SUI) by electrical stimulation, and the underlying mechanisms by which electrical stimulation regulates the collagen metabolism of female
Cheng Wang et al.
Oncology reports, 44(2), 711-721 (2020-07-07)
Mounting evidence indicates that there exists an association between heparanase (HPSE) and several physiological and pathological mechanisms in humans. However, the dynamics of the mechanisms involved in the regulation of HPSE expression in pancreatic cancer (PC) remain unclear. The aim
Yajun Wei et al.
Cancer management and research, 12, 2097-2108 (2020-04-08)
There is a growing number of evidence which report the relationship of the dual-specificity phosphatases 14 (DUSP14) with physiological and pathological mechanisms in the human body. However, it is still not known what if any role DUSP14 plays in pancreatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service