RFA451811K
Rotor for Eppendorf® 5424/5424R Centrifuges
FA-45-18-11-Kit, non aerosol tight, to hold, 18 x spin columns, lid, to hold, 18 x 1/5.2.0 mL tubes
Select a Size
$1,030.00
Estimated to ship onJune 02, 2025
Select a Size
About This Item
$1,030.00
Estimated to ship onJune 02, 2025
Recommended Products
feature
lid
to hold 18 x 1/5.2.0 mL tubes
to hold 18 x spin columns
fixed angle rotor with higher edge
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022653083 (NA)
Eppendorf® 5424706002 (EU)
parameter
18,111 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5424/5424R
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)