Skip to Content
MilliporeSigma

M3152

Methoxypolyethylene glycol succinate N-hydroxysuccinimide

~50%, mol wt ~5,000

Synonym(s):

mono-Methyl polyethylene glycol succinate N-succinimidyl ester, O-Methyl-O′-succinylpolyethylene glycol 5,000 N-succinimidyl ester, PEG-succinimidyl succinate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
PubChem Substance ID:
UNSPSC Code:
12161900
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mol wt

~5,000

concentration

~50%

storage temp.

−20°C

InChI

1S/C11H15NO7/c13-5-6-18-7-8(14)1-4-11(17)19-12-9(15)2-3-10(12)16/h13H,1-7H2

InChI key

OVPTZXRXNNJKSJ-UHFFFAOYSA-N

Application

For protein modification. May improve solubility and extend biological lifetime.

Other Notes

Shearwater Polymers product


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library