Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M1684

Sigma-Aldrich

(2S,3S,4S)-2-Methyl-2-(carboxycyclopropyl)glycine

Synonym(s):

(2S,3S,4S)-MCCG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO4
CAS Number:
Molecular Weight:
173.17
MDL number:
UNSPSC Code:
12352200

solubility

H2O: >10 mg/mL

storage temp.

−20°C

SMILES string

C[C@](N)([C@H]1C[C@@H]1C(O)=O)C(O)=O

Biochem/physiol Actions

Potent antagonist at mGluR2 metabotropic glutamate receptors

Analysis Note

Stereochemistry confirmed by x-ray crystallography

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Attwell et al.
Brain research, 805(1-2), 138-143 (1998-09-12)
The anticonvulsant effects of intracerebral administration of the highly potent group II metabotropic glutamate receptor agonist, DCG-IV, were tested in fully kindled rats following daily electrical stimulation of the basolateral amygdala. The agonist caused a dose-dependent increase in the generalized
T Knöpfel et al.
Neuropharmacology, 34(8), 1099-1102 (1995-08-01)
The two reported metabotropic glutamate receptor (mGluR) antagonists, alpha-methyl-cyclopropyl glycine (MCCG) and alpha-methyl-aminophosphonobutyrate (MAP4) were tested on the mGluR1b, mGluR2 and mGluR4a subtypes of human mGluRs. Neither MCCG (500 microM) nor MAP4 (500 microM) antagonized the activation of mGluR1b by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service