Skip to Content
MilliporeSigma
All Photos(1)

Documents

I5022

SAFC

Lithium ionophore III

Synonym(s):

ETH 1810, N,N-Dicyclohexyl-N′,N′-diisobutyl-cis-cyclohexane-1,2-dicarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H50N2O2
CAS Number:
Molecular Weight:
446.71
Beilstein/REAXYS Number:
6536266
MDL number:
UNSPSC Code:
12352200

storage temp.

2-8°C

SMILES string

CC(C)CN(CC(C)C)C(=O)[C@H]1CCCC[C@H]1C(=O)N(C2CCCCC2)C3CCCCC3

InChI

1S/C28H50N2O2/c1-21(2)19-29(20-22(3)4)27(31)25-17-11-12-18-26(25)28(32)30(23-13-7-5-8-14-23)24-15-9-6-10-16-24/h21-26H,5-20H2,1-4H3/t25-,26+/m0/s1

InChI key

QGKITQHJONXIQV-IZZNHLLZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Zeevi et al.
The Journal of membrane biology, 121(2), 133-140 (1991-04-01)
The neutral, noncyclic Li(+)-selective ionophore ETH1810, which is a di-imide, differs structurally from previous similar ionophores by removal of the intramolecular symmetry of the N-imide substituents. Properties of this ionophore, as a potential carrier of lithium, were probed through studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service