HUMANPBMC-0002201
Frozen Human Rheumatoid Arthritis PBMC
human peripheral blood (mononuclear cells)
Synonym(s):
Immune Cells, Peripheral Blood Mononuclear Cells, RA PBMC, Rheumatoid Arthritis PBMC
About This Item
Recommended Products
product name
Frozen Human Rheumatoid Arthritis PBMC, 50 Million Cells/per vial
biological source
human peripheral blood (mononuclear cells)
Quality Level
form
frozen liquid
packaging
vial of 50 Million cells
manufacturer/tradename
BioIVT
technique(s)
cell culture | mammalian: suitable
color
cloudy
storage temp.
-140 to -196°C
General description
We are pleased to offer these products from BioIVT.Offering isolated Peripheral Blood Mononuclear Cells(PBMCs) from whole blood procured from IRB-consented donors with clinicallydefined Rheumatoid Arthritis.
Peripheral Blood MononuclearCells (PBMCs) are a subset of leukocytes (or White Blood Cell, WBC), which areintegral components of the body′simmune system that fight disease.
PBMCs consist of a mixtureof lymphocytes (T cells, B cells, and NK cells) and monocytes and are isolatedfrom peripheral whole blood via density gradient centrifugation and red bloodcell lysis.
Standard and custom clinical data collection andconfirmation ensures proper corresponding disease severity and treatmenthistory (prescription and over the counter).
Clinical datamay include: Current medical treatment, CDAI Score, CRP Score, HLA, RF, CCPOurreadily available
PBMCs are provided with clinical data thathas been collected following a strict and standardized clinical data process.All specimens are reviewed for IRB approval.
Packaging
Other Notes
Disclaimer
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service