Documents
HS120768
Coated Wire Tube Rack
to hold, 80 x 20-25 mm tubes, blue
Synonym(s):
Tube holder, bottle rack
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
material
blue
high-density polyethylene coating
feature
to hold 80 x 20-25 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific 120768
L × W × H
11.4 in. (29 cm) × 9.1 in. (23 cm) × 3.1 in. (8 cm)
General description
The HDPE Coated Wire Rack is durable for transport or sample handling. It is chemically-resistant and suitable for conical, round or flat-bottom, or plastic tubes and bottles.
Features and Benefits
  • Chemically-resistant HDPE Coated Wire Racks
  • Racks can be used between temperatures of -40°C and 121°C
  • Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
  • Square wells and three deck levels help keep tubes secure
  • AutoclavableMesh Bottom Support