Documents
HS120622
MagFuge High Speed Centrifuge Rotor
to hold, 12 x 1.5/2 mL tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.78
feature
to hold 12 x 1.5/2 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120622
parameter
12,500 rpm speed (max.)
9,783 × g max. RCF
compatibility
Heathrow Scientific HS10050
Legal Information
MagFuge is a trademark of Heathrow Scientific